Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TFF3 Protein

Catalog No. 8294TF050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8294TF050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8294TF050 Supplier R&D Systems™ Supplier No. 8294TF050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce ERK1/ERK2 phosphorylation in Jurkat human acute T cell leukemia cells. 5-15 μg/mL of Recombinant Human TFF3 can effectively induce ERK1/2 phosphorylation.

Specifications

Accession Number Q07654
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7033
Molecular Weight (g/mol) M.W. (Observed): 7-11 kDa, reducing conditions; M.W. (theoretical): 7.4 kDa
Name TFF3
Preparation Method Reconstitute at 500μg/mL in PBS.
Quality Control Testing SDS-PAGE: 7-11kDa, reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human TFF3 protein Glu22-Phe80 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Glu22
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TFF3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TFF3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.