Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-alpha Protein

Catalog No. 239A500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
239A500 500 μg
239A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 239A500 Supplier R&D Systems™ Supplier No. 239A500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human TGF-alpha Protein is derived from E. coli. The Recombinant Human TGF-alpha Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 7039
Molecular Weight (g/mol) 6 kDa
Name TGF-alpha
Quantity 500 μg
Source E. coli-derived human TGF-alpha protein Val40-Ala89
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.