Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 (CHO-Expressed) Protein, CF
SDP

Catalog No. 11409BH010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-409-BH010 10 μg
11-409-BH500 500 μg
11-409-BH100 100 μg
11-409-BH01M 1 mg
11-409-BH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11-409-BH010 Supplier R&D Systems™ Supplier No. 11409BH010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.025-0.200 ng/mL.

Specifications

Accession Number P01137.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) M.W. (Observed): 12 kDa, under reducing conditions & 24 kDa under non-reducing conditions., M.W. (Theroretical): 13 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in 4 mM HCl. Reconstitute all other sizes at 250 μg/mL in 4 mM HCl.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TGF-beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.