Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein

Catalog No. 7754BH01M
Change view
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-54B-H01M 1000 μg
77-54B-H100 100 μg
77-54B-H500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 77-54B-H01M Supplier R&D Systems™ Supplier No. 7754BH01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human TGF-beta 1 (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P01137
For Use With (Application) Bioactivity
Gene ID (Entrez) 7040
Molecular Weight (g/mol) 12.8 kDa (monomer)
Quantity 1000 μg
Source Human embryonic kidney cell,HEK293-derived human TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Gene Symbol TGFB1
Specific Reactivity TGF-beta 1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 4 mM HCl containing at least 0.1% human or bovine serum albumin.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.