Learn More
Description
Specifications
Specifications
| Accession Number | NP_000651 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 10mM Sodium Citrate buffer (pH 3.5), 10% glycerol |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | 12.9kDa |
| Name | TGF-beta 1 Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MALDTNYCFS STEKNCCVRQ LYIDFRKDLG WKWIHEPKGY HANFCLGPCP YIWSLDTQYS KVLALYNQHN PGASAAPCCV PQALEPLPIV YYVGRKPKVE QLSNMIVRSC KCS |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
