Learn More
Creative Biomart Avotermin, Recombinant human TGF beta 3/TGFB3 Protein
Supplier:  Creative Biomart THP0318
Encompass_Preferred
It is a type of protein, known as a cytokine, which is involved in cell differentiation, embryogenesis and development. It belongs to a large family of cytokines called the Transforming growth factor beta superfamily, which includes the TGF- family, Bone morphogenetic proteins (BMPs), growth and differentiation factors (GDFs), inhibins and activins.
Catalog No. 50-208-4368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.