Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 3 (CHO-expressed) Protein

Catalog No. 8420B3025CF
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
84-20B-3025CF 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
84-20B-3005CF 5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
84-20B-3025 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein
84-20B-3005 5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 84-20B-3025CF Supplier R&D Systems™ Supplier No. 8420B3025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human TGF-beta 3 (CHO-expressed) Protein is derived from CHO. The Recombinant Human TGF-beta 3 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P10600
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
Gene ID (Entrez) 7043
Molecular Weight (g/mol) 13kDa
Name TGF-beta 3
Preparation Method Reconstitute at 50μg/mL in 4mM HCl.
Quality Control Testing SDS-PAGE: 9-11kDa, reducing conditions
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 3 protein Ala301-Ser412
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala301
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Gene Alias ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.