Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TGF-beta 3 Protein

Catalog No. 243B3002
Change view
Click to view available options
Quantity:
10 μg
2 μg
200 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
243B3002 2 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
243B3002CF 2 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
243B3010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
243B3010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
24-3B3-200 200 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
24-3B3-200CF 200 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 243B3002 Supplier R&D Systems™ Supplier No. 243B3002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.01-0.04 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 7043
Molecular Weight (g/mol) 12.7 kDa (monomer)
Name TGF-beta 3
Quantity 2 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TGF-beta 3 protein Ala301-Ser412 (Tyr340Phe)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.