Learn More
Creative Biomart GMP Grade Recombinant Human TGFB3 protein
Supplier:  Creative Biomart TGFB3193THP
Encompass_Preferred
Recombinant Human TGFB3 protein GMP Grade(NP_003230)(375 aa) was expressed in HEK293 cells with a C-terminal Fc Tag.
Catalog No. 50-208-5254
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.