Learn More
R&D Systems™ Recombinant Human Thioredoxin 80 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris, EDTA and DTT. |
| Gene ID (Entrez) | 7295 |
| Molecular Weight (g/mol) | 9 kDa (monomer) |
| Name | Thioredoxin-80 |
| Quantity | 100 μg |
| Source | E. coli-derived human Thioredoxin-80 protein Val2-Lys81 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homodimer |
| Endotoxin Concentration | <0.50 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.