Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human THOC7 His Protein
SDP

Catalog No. NBP25195005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25195005 0.05 mg
NBP25195025 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25195005 Supplier Novus Biologicals™ Supplier No. NBP2519500.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-204 of Human THOC7 The Recombinant Human THOC7 Protein is derived from E. coli. The Recombinant Human THOC7 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing, 50% glycerol, 1mM DTT
Gene ID (Entrez) 80145
Molecular Weight (g/mol) TMW: 26.1kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias FLJ23445, fSAP24NIF3L1-binding protein 1, Functional spliceosome-associated protein 24, Ngg1 interacting factor 3 like 1 binding protein 1, Ngg1-interacting factor 3-like protein 1-binding protein 1, NIF3L1BP1hTREX30, THO complex 7 homolog (Drosophila), THO complex subunit 7 homolog
Common Name THOC7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.