Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Thrombopoietin (E. coli-expressed)

Catalog No. 288TPE100
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
288TPE100 100 μg
28-8TP-E010 10 μg
28-8TP-E050 50 μg
288TPE01M 1 mg
288TPE250 250 μg
288TPE500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 288TPE100 Supplier R&D Systems™ Supplier No. 288TPE100
Add to Cart
Add to Cart
This item is not returnable. View return policy

A key regulator of megakaryocytopoiesis and thrombopoiesis

The Recombinant Human Thrombopoietin (E. coli-expressed) is derived from E. coli. The Recombinant Human Thrombopoietin (E. coli-expressed) has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_000451
For Use With (Application) Bioactivity
Gene ID (Entrez) 7066
Molecular Weight (g/mol) 18.7 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 100 μg
Source E. coli-derived human Thrombopoietin/Tpo protein Ser22-Leu195
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 ug of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 ug/mL in PBS
Buffer Lyophilized from a 0.2 um filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.