Learn More
R&D Systems™ Recombinant Human Thrombopoietin (E. coli-expressed)
Description
Specifications
Specifications
| Accession Number | NP_000451 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 7066 |
| Molecular Weight (g/mol) | 18.7 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 500 μg |
| Source | E. coli-derived human Thrombopoietin/Tpo protein Ser22-Leu195 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 ug of the protein by the LAL method |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.