Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Thrombopoietin/THPO Protein
SDP

Catalog No. NBP275205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275205 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275205 Supplier Novus Biologicals™ Supplier No. NBP275205

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein for Human Thrombopoietin/Tpo The Recombinant Human Thrombopoietin/THPO Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration LYOPH
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized with no additives
Gene ID (Entrez) 7066
Quantity 0.01 mg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Symbol THPO
Cross Reactivity Thrombopoietin/THPO
Species Human
Reconstitution Reconstitution in purified H2O to a concentration of 0.1-1.0 mg/ml. The solution can then be diluted into other aqueous buffers and stored at 4C for 1 week or -20C for long-term storage. For long term storage add a carrier protein (0.1% HAS or BSA).
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.