Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Tie-2 His-tag Protein

Catalog No. 10457TI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10457TI050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10457TI050 Supplier R&D Systems™ Supplier No. 10457TI050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Tie-2 His-tag (Catalog # 10457-TI) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Human Angiopoietin-2 His-tag (Catalog # 623-AN ) binds with an ED50 of 0.5-4 ng/mL.

Specifications

Accession Number AAA61139.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7010
Molecular Weight (g/mol) M.W.-observed: 98-111 kDa, under reducing conditions, M.W.-theoretical: 81 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Tie-2 protein Ala23-Lys745, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Tie-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.