Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TINAGL1 Protein

Catalog No. 9304TU050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-04T-U050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-04T-U050 Supplier R&D Systems™ Supplier No. 9304TU050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human TINAGL1 Protein is derived from CHO. The Recombinant Human TINAGL1 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature

Specifications

Accession Number Q9GZM7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl and Brij-35
Gene ID (Entrez) 64129
Molecular Weight (g/mol) 52 kDa
Name TINAGL1 Recombinant Protein
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TINAGL1 protein Human TINAGL1 (Ala22-Met464) Accession &Num; Q9GZM7 HP GGGSGGGSGGGS HHHHHH
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Common Name androgen-regulated gene 1; ARG1; GIS5; Glucocorticoid-inducible protein 5; LCN7; LIECG3; lipocalin 7; OLRG2; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; TIN Ag-related protein; TINAGL; TINAGL1; TINAG-like 1; TIN
Biological Activity 1 to 6ng/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.