Learn More
Description
Specifications
Specifications
| Accession Number | Q9NYK1 |
| For Use With (Application) | Flow Cytometry |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 51284 |
| Molecular Weight (g/mol) | 120 kDa |
| Name | toll-like receptor 7 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human TLR7 protein, (N-terminus) Human TLR7 (Ala27-Asn839) Accession &Num; Q9NYK1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
