Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 54664 |
| Molecular Weight (g/mol) | M.W.-observed: >60 kDa, under reducing conditions., M.W.-theroretical: 45 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human TMEM106B protein Human IgG1 (Pro100-Lys330) IEGR Human TREM106B (Pro118-Gln274) Accession # (N-terminus)NP_001127704.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | transmembrane protein 106B |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
