Learn More
R&D Systems™ Recombinant Human TNF-alpha Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | P01375.1 |
| For Use With (Application) | Bioactivity, Gel Filtration Chromatography |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 7124 |
| Molecular Weight (g/mol) | M.W.-observed: 15-22 kDa, under reducing conditions, M.W.-theroretical: 19 kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TNF-alpha protein Avi-tag Human TNF alpha (Val77-Leu233) (c-terminus) Accession # P01375.1 N-terminusC-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.