Learn More
R&D Systems™ Recombinant Human TNF-alpha (HEK293-expressed) Protein
Specifications
Specifications
| Accession Number | P01375 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 7124 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Quantity | 250 μg |
| Source | Human embryonic kidney cell, HEK293-derived human TNF-alpha protein Val77-Leu233 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.