Learn More
R&D Systems™ Recombinant Human TNF RII/TNFRSF1B Fc Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | AAA36755.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 7133 |
| Molecular Weight (g/mol) | M.W.-observed: 68-81 kDa, under reducing conditions, M.W.-theroretical: 54 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human TNF RII/TNFRSF1B protein Human TNF RII/TNFRSF1B (Leu23-Asp257) (N-terminus) Accession #AAA36755.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1 B, tumor necrosis factor receptor superfamily, member 1 B, Tumor necrosis factor receptor type II |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.