Learn More
R&D Systems™ Recombinant Human TRAIL R2 Fc Alexa Fluor™ 647 Protein
Description
Specifications
Specifications
| Accession Number | AAH01281.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8795 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 54-61 kDa, under reducing conditions., MolecularWeight-theroretical: 44 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TRAIL R2/TNFRSF10B protein Human TRAIL R2/TNFRSF10B (Ala54-Ser210) (N-terminus) Accession # AAH01281.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2 L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2 B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10 B, tumor necrosis factor receptor superfamily, member 10 b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.