Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein

Catalog No. 375TEC010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 .
Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
375TEC010 Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 with BSA as a carrier protein.
375TEC010CF Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 .
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 375TEC010 Supplier R&D Systems™ Supplier No. 375TEC010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein is derived from E. coli. The Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 with BSA as a carrier protein.
Gene ID (Entrez) 8743
Molecular Weight (g/mol) 19 kDa (monomer)
Name TRAIL/TNFSF10
Quantity 10 μg
Source E. coli-derived human TRAIL/TNFSF10 protein Val114-Gly281,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 2 weeks, 2 to 8° C under sterile conditions after reconstitution. 1 month, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Homotrimer
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.