Learn More
R&D Systems™ Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 . |
| Gene ID (Entrez) | 8743 |
| Molecular Weight (g/mol) | 19 kDa (monomer) |
| Name | TRAIL/TNFSF10 |
| Quantity | 10 μg |
| Source | E. coli-derived human TRAIL/TNFSF10 protein Val114-Gly281,with and without an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 2 weeks, 2 to 8° C under sterile conditions after reconstitution. 1 month, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Homotrimer |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.