Learn More
R&D Systems™ Recombinant Human TRANCE/RANK L His Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | O14788.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 8600 |
| Molecular Weight (g/mol) | M.W.-observed: 26-39 kDa under reducing conditions., M.W.-theroretical: 24 kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TRANCE/TNFSF11/RANK L protein Avi-tag HHHHHH Human TRANCe/TNFSF11/RANK L (Ile140-Asp317) (N-terminus) Accession # O14788.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.