Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Transglutaminase 2/TGM2 Protein

Catalog No. 4376TG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4376TG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4376TG050 Supplier R&D Systems™ Supplier No. 4376TG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Transglutaminase 2/TGM2 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Transglutaminase 2/TGM2 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT.
Gene ID (Entrez) 7052
Molecular Weight (g/mol) 78 kDa
Name Transglutaminase 2/TGM2
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Transglutaminase 2/TGM2 protein Ala2-Ala687 (Asn533Thr and Leu655Val),with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.