Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human Transthyretin/Prealbumin Y78F Variant, Monomer Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Specifications
Specifications
For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
Formulation | PBS pH 7.4 |
Gene ID (Entrez) | 7276 |
Quantity | 100 μg |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Gene Alias | ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin |
Gene Symbol | TTR |
Research Category | Cancer |
Purity or Quality Grade | >95%, by SDS-PAGE |
Protein | Transthyretin/Prealbumin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction