Learn More
R&D Systems™ Recombinant Human TrkA Fc Chimera Avi-tag Protein
Description
When Recombinant Human beta-NGF (Catalog # 256-GF ) is immobilized at 0.5 μg/mL (100 μL/well), Biotinylated Recombinant Human TrkA Fc Chimera Avi-tag (Catalog # AVI11378) binds with an ED50 of 7.00-70.0 ng/mL.
Specifications
Specifications
| Accession Number | P04629.4 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4914 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 105-120 kDa, under reducing conditions., MolecularWeight-theroretical: 71 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TrkA protein Human TrkA (Ala34-Gly423) (N-terminus) Accession # P04629.4 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.