Learn More
Creative Biomart Recombinant Human Tropoelastin
Supplier:  Creative Biomart THP0321
Encompass_Preferred
Recombinant Human Tropoelastin is the precursor to elastin, a structural protein made by crosslinking together the water soluble tropoelastin molecules. Elastin provides strength and elasticity and is the main constituent of elastic connective tissue. It allows the tissue to resume its shape after contracting or stretching and is found in the skin, heart, blood vessels, lungs, ligaments, and the bladder.Tropoelastin is supplied as a recombinant lyophilized protein and can be used to coat cell culture surfaces to enhance cell attachment and promote spreading.
Catalog No. 50-208-4371
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.