Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TSPAN7/TM4SF2 Protein
SDP

Catalog No. NBP25957550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25957550 50 μg
NBP259575Y 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25957550 Supplier Novus Biologicals™ Supplier No. NBP25957550UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 113-213 of Human TSPAN7/TM4SF2 The Recombinant Human TSPAN7/TM4SF2 Protein is derived from E. coli. The Recombinant Human TSPAN7/TM4SF2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 7102
Molecular Weight (g/mol) 12.6 kDa
Name Recombinant Human TSPAN7/TM4SF2 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-ass
Gene Symbol TSPAN7
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.