Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 5328 |
| Molecular Weight (g/mol) | 18 kDa (A chain), 30 kDa (B chain) |
| Name | u-Plasminogen Activator/Urokinase |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human u-Plasminogen Activator (uPA)/Urokinase protein Met1-Leu431 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Gene Alias | ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, u-PA, UPA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
