Learn More
R&D Systems™ Recombinant Human Ubiquitin+1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 13 kDa |
| Name | Ubiquitin+1 |
| Quantity | 25 μg |
| Source | E. coli-derived human Ubiquitin+1 protein ATVID 10-His tag SS Ubiquitin (Met1 – Gly75) Accession &Num; CAA44911 YADLREDPDRQDHHPGSGAQ N-terminus C-terminus The italicized carboxyl terminal sequence is generated by a frameshift in the mRNA. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.