Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Ubiquitin Activating Enzyme (UBE1)

Catalog No. E305025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E305025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E305025 Supplier R&D Systems™ Supplier No. E305025/DISC
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Recombinant Human Ubiquitin Activating Enzyme (UBE1) is a member of the Ubiquitin-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a Ubiquitin-conjugating (E2) enzyme and a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human Ubiquitin Activating Enzyme (UBE1) concentration of 50-200 nM.

Specifications

Accession Number P22314.3
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl and TCEP.
Gene ID (Entrez) 7317
Molecular Weight (g/mol) M.W. (theoretical): 118 kDa
Name Ubiquitin-activating Enzyme/UBE1
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Ubiquitin-activating Enzyme/UBE1 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol UBA1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Ubiquitin-activating Enzyme/UBE1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.