Learn More
R&D Systems™ Recombinant Human Ubiquitin Biotin Protein, CF
Description
- Derived from E. coli
- Validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P0CG47 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 6233 |
| Quantity | 100 μg |
| Source | E. coli-derived human Ubiquitin protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -20 to -70 °C as supplied. 12 months, -20 to -70 °C under sterile conditions after opening. |
| Conjugate | Biotin |
| Species | Human |
| Buffer | X mg/mL in 10 mm HEPES pH 7.5 |
| Purity or Quality Grade | > 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.