Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Ubiquitin N-Terminal Fluorescein

Catalog No. U580050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U580050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. U580050 Supplier R&D Systems™ Supplier No. U580050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Ubiquitin N-Terminal Fluorescein is derived from E. coli. The Recombinant Human Ubiquitin N-Terminal Fluorescein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P0CG47.1
Formulation Lyophilized from a solution in deionized water.
Gene ID (Entrez) 7314
Molecular Weight (g/mol) M.W. Theroretical: 9 kDa
Name Ubiquitin N-Terminal Fluorescein
Quantity 50 μg
Source E. coli-derived human Ubiquitin protein Gln2 - Gly76 with a single N-terminal fluorescein
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Conjugate Fluorescein
Recombinant Recombinant
Reconstitution Reconstitute in aqueous buffer or DMSO to 1mg/mL
Purity or Quality Grade 95%, by PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.