Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Ubiquitin Rhodamine 110 Protein

Catalog No. U555050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U555050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. U555050 Supplier R&D Systems™ Supplier No. U555050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Ubiquitin Rhodamine 110 Protein is derived from E. coli. The Recombinant Human Ubiquitin Rhodamine 110 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a solution in DMSO.
Gene ID (Entrez) 7314
Molecular Weight (g/mol) 8.9 kDa
Name Ubiquitin Rhodamine 110 C-Terminal Derivative
Quantity 50 μg
Source E. coli-derived human Ubiquitin protein Contains a C-terminal Rhodamine 110 (R110)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate R110
Recombinant Recombinant
Purity or Quality Grade 95%, by HPLC.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.