Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ULBP-1 Fc Chimera Protein

Catalog No. 1380UL01M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-80U-L01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-80U-L01M Supplier R&D Systems™ Supplier No. 1380UL01M
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human ULBP-1 Fc Chimera Protein is derived from Sf 21 (stably transfected). The Recombinant Human ULBP-1 Fc Chimera Protein has been validated for the following applications: Binding Activity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q9BZM6
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2μ filtered solution in PBS
Gene ID (Entrez) 80329
Molecular Weight (g/mol) 48.8 kDa (monomer)
Quantity 1 mg
Source Spodoptera frugiperda,Sf 21 (stably transfected)-derived human ULBP-1 protein, (N-terminus) Human ULBP-1 (Gly26-Pro215) Accession &Num; Q9BZM6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1
Gene Symbol ULBP1
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.