Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ULBP-2 His-tag Protein

Catalog No. 11196UL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-196-UL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-196-UL050 Supplier R&D Systems™ Supplier No. 11196UL050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Recombinant Human NKG2D/CD314 Fc Chimera (Catalog # 1299-NK ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human ULBP-2 Hs--tag (Catalog # 11196-UL) binds with an ED50 of 7.50-60.0 ng/mL.

Specifications

Accession Number Q9BZM5.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 80328
Molecular Weight (g/mol) M.W. Observed: 25-35 kDa, under reducing conditions., M.W. Theoretical: 23 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ULBP-2 protein Gly26-Ser217, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, NKG2DL2, RAET1H, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2, ULBP2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ULBP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.