Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human UNC5B Protein Fc Chimera
SDP

Catalog No. 8869UN050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-69U-N050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 88-69U-N050 Supplier R&D Systems™ Supplier No. 8869UN050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human UNC5B Protein Fc Chimera is derived from CHO. The Recombinant Human UNC5B Protein Fc Chimera has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8IZJ1
For Use With (Application) Bioactivity
Gene ID (Entrez) 219699
Molecular Weight (g/mol) Theoretical M.W.: 66kDa (monomer)
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human UNC5H2/UNC5B protein, (N-terminus) Human UNC5H2/UNC5B (Gly27-Tyr377) Accession &Num; Q8IZJ1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias p53RDL1, p53-regulated receptor for death and life protein 1, Protein unc-5 homolog 2, Protein unc-5 homolog B, transmembrane receptor Unc5H2, unc5 (C.elegans homolog) b, unc-5 homolog 2, unc-5 homolog B (C. elegans), UNC5B, UNC5H2netrin receptor UNC5B
Common Name UNC5H2/UNC5B
Gene Symbol UNC5B
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human UNC5H2/UNC5B Fc Chimera is immobilized at 2μg/mL (100μL/well), the concentration of Recombinant Mouse Netrin-1 that produces 50% optimal binding response is approximately 7-42ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human UNC5H2/UNC5B (Gly27-Tyr377) Accession # Q8IZJ1, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.