Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human VAMP3/Cellubrevin Protein
SDP

Catalog No. NBC118346 Shop All R&D Systems Products
Change view
Click to view available options
:
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBC118346 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118346 Supplier Novus Biologicals™ Supplier No. NBC118346
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-77 of Human VAMP3/Cellubrevin The Recombinant Human VAMP3/Cellubrevin Protein is derived from E. coli. The Recombinant Human VAMP3/Cellubrevin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004772
For Use With (Application) Western Blot
Formulation 20mM Tris-HCl buffer (pH 7.5), 10% glycerol
Gene ID (Entrez) 9341
Molecular Weight (g/mol) 8.7kDa
Name VAMP3/Cellubrevin Protein
Purification Method Protein
Immunogen MSTGPTAATGSNRRLQQTQNQVDEVVDIMRVNVDKVLERDQKLSELDDRADALQAGASQFETSAAKLKRKYWWKNCK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.