Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Vasorin/SLIT-like 2 Fc Chimera Protein
SDP

Catalog No. 10139VN050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-139-VN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-139-VN050 Supplier R&D Systems™ Supplier No. 10139VN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Vasorin, also known as SLIT-like 2, is an approximately 110-kDa transmembrane protein that is expressed on vascular smooth muscle cells and in the developing skeletal system

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number Q6EMK4
For Use With (Application) Bioactivity
Gene ID (Entrez) 114990
Molecular Weight (g/mol) 86 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human Vasorin/SLIT-like 2 protein

Human Vasorin/SLIT-like 2 (Cys24-Asn573) Accession # Q6EMK4IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.