Learn More
R&D Systems™ Recombinant Human VEGF 165, ACFP Protein
Description
- ED50 for this effect is 1 to 6 ng/mL.
- Specific activity of recombinant human VEGF is approximately 1.7 x 103 U/μg, which is calibrated against recombinant human VEGF 165 WHO Standard (NIBSC code: 02/286).
Specifications
Specifications
| Accession Number | NP_001165097 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | 19 to 21 kDa, reducing conditions |
| Quantity | 1 mg |
| Source | Spodoptera frugiperda, Sf 9 (baculovirus)-derived human VEGF protein Met1-Arg191 Produced in an animal component free process (ACFP) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.01 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.