Learn More
R&D Systems™ Recombinant Human VEGF 165 (E. coli-expressed) Protein, CF
Description
Specifications
Specifications
| Accession Number | NP_001165097 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in HCl. |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | observed_molecular_weight: 19-22 kDa, under reducing conditions. |
| Quantity | 10 μg |
| Source | E. coli-derived human VEGF protein Ala27-Arg191 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 100 μg/mL in sterile PBS. Alternatively, reconstitute (at 100-500 μg/mL in sterile 4 mM HCl..) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.