Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 (HEK293-expressed) Protein, CF

Catalog No. 11458VE01M
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-458-VE01M 1 mg
11-458-VE500 500 μg
11-458-VE250 250 μg
11-458-VE100 100 μg
11-458-VE050 50 μg
11-458-VE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 11-458-VE01M Supplier R&D Systems™ Supplier No. 11458VE01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.

Specifications

Accession Number NP_001165097.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) MolecularWeight-observed: 19-25 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in 4mM HCl. Reconstitute all other sizes at 500 μg/mL in 4mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VEGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.