Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein
GSA_VASDP

Catalog No. 293VE010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
293VE010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
293VE050 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
293VE010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
293VE001MGC 1 mg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
293VE050CF 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
29-3VE-500CF 500 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
29-3VE-500 500 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 293VE010 Supplier R&D Systems™ Supplier No. 293VE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1-6 ng/mL. The specific activity of recombinant human VEGF165 is approximately 1.7 x 103 U/μg, which is calibrated against recombinant human VEGF165 WHO Standard (NIBSC code: 02/286). Specific activity is for reference purposes only and is not routinely tested.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) observed_molecular_weight: 20-22 kDa, under reducing conditions. 39-42 kDa, under non-reducing conditions.
Name VEGF 165
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. Alternatively, reconstitute (at 100-500 μg/mL in sterile 4 mM HCl containing 0.1% human or bovine serum albumin.)
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.