Learn More
Description
Specifications
Specifications
| Accession Number | NP_001020539 |
| Concentration | 0.5mg/mL |
| Formulation | 20mM Tris-HCl(pH8.5), 50% glycerol, 5mM DTT, 200mM NaCl, 2mM EDTA |
| Gene ID (Entrez) | 7422 |
| Quantity | 100 μg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Species | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
