Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein

Catalog No. 293VE01M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-3VE-01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-3VE-01M Supplier R&D Systems™ Supplier No. 293VE01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1-6 ng/mL. The specific activity of recombinant human VEGF165 is approximately 1.7 x 103 U/μg, which is calibrated against recombinant human VEGF165 WHO Standard (NIBSC code: 02/286). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number NP_001165097
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) observed_molecular_weight: 20-22 kDa, under reducing conditions. 39-42 kDa, under non-reducing conditions.
Quantity 1 mg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01EU per 1μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Gene Symbol VEGFA
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. Alternatively, reconstitute (at 100-500 μg/mL in sterile 4 mM HCl containing 0.1% human or bovine serum albumin.)
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.