Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF-Ax Protein
SDP

Catalog No. 9018VE025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-18V-E025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-18V-E025 Supplier R&D Systems™ Supplier No. 9018VE025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human VEGF-Ax Protein is derived from CHO. The Recombinant Human VEGF-Ax Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_001303939
For Use With (Application) Bioactivity
Gene ID (Entrez) 7422
Molecular Weight (g/mol) Observed M.W.: 22-29kDa, reducing conditions, Theoretical M.W.: 21kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human VEGF protein, (N-terminus) Human VEGF-A (Ala27-Arg191) Accession &Num; NP_001303939 SAGQEEGASLRVSGTRSLTRKD (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Common Name VEGF
Gene Symbol VEGFA
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R2/KDR/Flk-1 Fc Chimera is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human VEGF-Ax that produces 50% of the optimal binding response is approximately 1-6ng/mL. Recombinant Human VEGF-Ax weakly binds to Recombinant Human Neuropilin-1.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human VEGF-A (Ala27-Arg191) Accession # NP_001303939, N-terminus; SAGQEEGASLRVSGTRSLTRKD, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.