Learn More
R&D Systems™ Recombinant Human Versican Isoform V3 His-tag Protein, CF
Description
- Recombinant Protein
- Carrier Free (CF)
- The carrier free version does not contain Bovine Serum Albumin (BSA)
Specifications
Specifications
| Accession Number | P13611-4 |
| Gene ID (Entrez) | 1462 |
| Molecular Weight (g/mol) | 71 kDa |
| Purification Method | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Versican protein Leu21-Tyr646, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.