Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VSIG4 Short Isoform Fc Chimera Protein

Catalog No. 9817VS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-17V-S050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-17V-S050 Supplier R&D Systems™ Supplier No. 9817VS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit anti-CD3 antibody induced IFN-gamma secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 1-5 μg/mL.

Specifications

Accession Number Q9Y279-3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11326
Molecular Weight (g/mol) M.W. (Observed): 55-65 kDa, reducing conditions; M.W. (theoretical): 46 kDa
Name CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human VSIG4 protein Human VSIG4 (Arg20-Pro189) (N-terminus) Accession # Q9Y279-3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VSIG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.